Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATP12A Partial ORF (NP_001667, 174 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000479Q01L

 View more versions of this product

Catalog No. 89-935-990


Add to cart

Description

Description

The protein encoded by this gene belongs to the family of P-type cation transport ATPases. This gene encodes a catalytic subunit of the ouabain-sensitive H+/K+ -ATPase that catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. It is also responsible for potassium absorption in various tissues. [provided by RefSeq]

Sequence: SSFNKMIPQQALVIRDSEKKTIPSEQLVVGDIVEVKGGDQIPADIRVLSSQGCRVDNSSLTGESEPQPRSSEFTHENPLETKNICFYSTTCLEASTSPVGTVTGMVIN
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SSFNKMIPQQALVIRDSEKKTIPSEQLVVGDIVEVKGGDQIPADIRVLSSQGCRVDNSSLTGESEPQPRSSEFTHENPLETKNICFYSTTCLEASTSPVGTVTGMVIN
RUO
ATP12A
Wheat Germ (in vitro)
GST
NP_001667
Liquid
479
ATP12A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP1AL1
ATP12A
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit