Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ATP6V1C1 Full-length ORF (NP_001686.1, 1 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000528P01S

 View more versions of this product

Catalog No. 89-936-075


Add to cart

Description

Description

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D. [provided by RefSeq]

Sequence: MTEFWLISAPGEKTCQQTWEKLHAATSKNNNLAVTSKFNIPDLKVGTLDVLVGLSDELAKLDAFVEGVVKKVAQYMADVLEDSKDKVQENLLANGVDLVTYITRFQWDMAKYPIKQSLKNISEIIAKGVTQIDNDLKSRASAYNNLKGNLQNLERKNAGSLLTRSLAEIVKKDDFVLDSEYLVTLLVVVPKLNHNDWIKQYETLAEMVVPRSSNVLSEDQDSYLCNVTLFRKAVDDFRHKARENKFIVRDFQYNEEEMKADKEEMNRLSTDKKKQFGPLVRWLKVNFSEAFIAWIHVKALRVFVESVLRYGLPVNFQAMLLQPNKKTLKKLREVLHELYKHLDSSAAAIIDAPMDIPGLNLSQQEYYPYVYYKIDCNLLEFK
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ATP6V1C1
Wheat Germ (in vitro)
GST
NP_001686.1
Liquid
528
ATP6V1C1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6C/ATP6D/FLJ20057/VATC/Vma5
ATP6V1C1
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit