Learn More
Abnova™ Human AURKC (NM_003160, 1 a.a. - 275 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4653
Catalog No. 89-936-178
Add to cart
Description
Description
This gene encodes a member of the Aurora subfamily of serine/threonine protein kinases. The encoded protein is a chromosomal passenger protein that forms complexes with Aurora-B and inner centromere proteins and may play a role in organizing microtubules in relation to centrosome/spindle function during mitosis. This gene is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Specifications
Specifications
Insect cell (Sf9) expression system
0.116 ug/uL
Liquid
6795
AURKC (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
AURKC
4pmol/μg x min
Yes
NM_003160
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
In 50mM Tris-HCl, 100mM NaCl, pH 8.0. (5mM DTT, 4mM reduced glutathione, 20% glycerol)
58.480kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
AIE2/AIK3/AurC/STK13/aurora-C
AURKC
Insect
GST-His