Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human AZI2 Full-length ORF (NP_071906.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00064343P01S
Description
AZI2, or NAP1, contributes to the activation of NFKB (see MIM 164011)-dependent gene expression by activating IKK-related kinases, such as NAK (TBK1; MIM 604834) (Fujita et al., 2003 [PubMed 14560022]).[supplied by OMIM]
Sequence: MDALVEDDICILNHEKAHKRDTVTPVSIYSGDESVASHFALVTAYEDIKKRLKDSEKENSLLKKRIRFLEEKLIARFEEETSSVGREQVNKAYHAYREVCIDRDNLKSKLDKMNKDNSESLKVLNEQLQSKEVELLQLRTEVETQQVMRNLNPPSSNWEVEKLSCDLKIHGLEQELELMRKECSDLKIELQKAKQTDPYQEDNLKSRDLQKLSISSDNMQHAYWELKREMSNLHLVTQVQAELLRKLKTSTAIKKACAPVGCSEDLGRDSTKLHLMNFTATYTRHPPLLPNGKALCHTTSSPLPGDVKVLSEKAILQSWTDNERSIPNDGTCFQEHSSYGRNSLEDNSWVFPSPPKSSETAFGETKTKTLPLPNLPPLHYLDQHNQNCLYKNSpecifications
NP_071906.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AZI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64343 | |
AZI2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZ2/NAP1/TILP | |
AZI2 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction