Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human AZIN1 Partial ORF (NP_056962, 339 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051582Q01L

 View more versions of this product

Catalog No. 89-936-228


Add to cart

Description

Description

Ornithine decarboxylase (ODC) catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. Ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein encoded by this gene is highly similar to ODC. It binds to ODC antizyme and stabilizes ODC, thus inhibiting antizyme-mediated ODC degradation. Two alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: HKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAE
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAE
RUO
AZIN1
Wheat Germ (in vitro)
GST
NP_056962
Liquid
51582
AZIN1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC3832/MGC691/OAZI/OAZIN/ODC1L
AZIN1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit