Learn More
R&D Systems™ Recombinant Human B7-1/CD80 His-tag Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9050B1100
Catalog No. 90-50B-1100
Add to cart
This item is not returnable. View return policy
Description
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human B7-1/CD80 is immobilized at 1 μg/mL (100 μL/well), the concentration of Recombinant Human CTLA-4 Fc Chimera (Catalog # 7268-CT ) that produces 50% of the optimal binding response is approximately 0.05-0.3 μg/mL.
Specifications
Specifications
P33681
Lyophilized from a 0.2 µm filtered solution in PBS.
M.W. (Observed): 45-58 kDa, reducing conditions; M.W. (theoretical): 25 kDa
100 µg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
CD80
Unconjugated
Yes
Human embryonic kidney cell, HEK293-derived Val35-Asn242, with a C-terminal 6-His tag
Bioactivity
941
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Human embryonic kidney cell, HEK293-derived human B7-1/CD80 protein Val35-Asn242, with a C-terminal 6-His tag
<0.10 EU / 1 µg of the protein by the LAL method.
B7-1/CD80
Measured by its binding ability in a functional ELISA. When Recombinant Human B7-1/CD80 is immobilized at 1μg/mL (100μL/well), the concentration of Recombinant Human CTLA-4 Fc Chimera that produces 50% of the optimal binding response is approximately 0.05-0.3μg/mL.
Human
Reconstitute at 500 µg/mL in PBS.
B7-1/CD80