Learn More
Raybiotech IncSupplier Diversity Partner Human B7-H3/CD276 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHB7H31
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 3.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human B7-H3.
Catalog No. 50-194-9011