Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BAG2 Full-length ORF (NP_004273.1, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name BAG2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 50.2kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-936-339
View Documents
Abnova™
H00009532P01L
25 ug
Each for $709.50
Add to cart
 
89-936-338
View Documents
Abnova™
H00009532P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner. [provided by RefSeq]

Sequence: MAQAKINAKANEGRFCRSSSMADRSSRLLESLDQLELRVEALREAATAVEQEKEILLEMIHSIQNSQDMRQISDGEREELNLTANRLMGRTLTVEVSVETIRNPQQQESLKHATRIIDEVVNKFLDDLGNAKSHLMSLYSACSSEVPHGPVDQKFQSIVIGCALEDQKKIKRRLETLLRNIENSDKAIKLLEHSKGAGSKTLQQNAESRFN
Specifications

Specifications

BAG2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
50.2kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit