Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human BAG4 Full-length ORF (AAH38505, 1 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $714.00

Specifications

Name BAG4 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 76.01kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-936-342
View Documents
Abnova™
H00009530P01S
10 ug
Each for $460.50
Add to cart
 
89-936-343
View Documents
Abnova™
H00009530P01L
25 ug
Each for $714.00
Add to cart
 
Description

Description

The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70 kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. This protein was found to be associated with the death domain of tumor necrosis factor receptor type 1 (TNF-R1) and death receptor-3 (DR3), and thereby negatively regulates downstream cell death signaling. The regulatory role of this protein in cell death was demonstrated in epithelial cells which undergo apoptosis while integrin mediated matrix contacts are lost. [provided by RefSeq]

Sequence: MSALRRSGYGPSDGPSYGRYYGPGGGDVPVHPPPPLYPLRPEPPQPPISWRVRGGGPAETTWLGEGGGGDGYYPSGGAWPEPGRAGGSHQEQPPYPSYNSNYWNSTARSRAPYPSTYPVRPELQGQSLNSYTNGAYGPTYPPGPGANTASYSGAYYAPGYTQTSYSTEVPSTYRSSGNSPTPVSRWIYPQQDCQTEAPPLRGQVPGYPPSQNPGMTLPHYPYGDGNRSVPQSGPTVRPQEDAWASPGAYGMGGRYPWPSSAPSAPPGNLYMTESTSPWPSSGSPQSPPSPPVQQPKDSSYPYSQSDQSMNRHNFPCSVHQYESSGTVNNDDSDLLDSQVQYSAEPQLYGNATSDHPNNQDQSSSLPEECVPSDESTPPSIKKIIHVLEKVQYLEQEVEEFVGKKTDKAYWLLEEMLTKELLELDSVETGGQDSVRQARKEAVCKIQAILEKLEKKGL
Specifications

Specifications

BAG4 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
76.01kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit