Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BAI1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4969
This item is not returnable.
View return policy
Description
BAI1 Monoclonal specifically detects BAI1 in Human samples. It is validated for Immunohistochemistry.Specifications
BAI1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
O14514 | |
BAI1 | |
Chinese hamster ovary cell line CHO-derived recombinant human BAI1 Met1-Thr879 Accession # O14514 | |
100 μg | |
Primary | |
Detects human BAI1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human BAI3 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
542515 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
575 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction