Learn More
R&D Systems™ Human BAI2 Fc Chimera Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9338BA050
Catalog No. 93-38B-A050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human BAI2 Protein Fc Chimera is derived from HEK293. The Recombinant Human BAI2 Protein Fc Chimera has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
O60241-3
576
>85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
HEK293
RUO
<0.10 EU per 1μg of the protein by the LAL method
Recombinant protein
Yes
Lyophilized from a 0.2μm filtered solution in PBS
123 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Disulfide-linked homodimer
25 to 150ng/mL
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.