Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human BCAM/CD239 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol BCAM
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB59644250U View Documents Novus Biologicals™
NBP259644250UG
250μg Each for $1,025.00
Add to cart
 
NB5964450UG View Documents Novus Biologicals™
NBP25964450UG
50μg Each for $287.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 32-547 of Human BCAM/CD239 The Recombinant Human BCAM/CD239 Protein is derived from Baculovirus. The Recombinant Human BCAM/CD239 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein
RUO
BCAM
In PBS (pH 7.4) containing 10% glycerol
E.coli
Unlabeled
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
antigen identified by monoclonal F8, Auberger B antigen
Human
AC
83.2 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit