Learn More
Novus Biologicals™ Human Bcl-2 related protein A1 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269948
Catalog No. NBP269948
Add to Cart
Description
Description
The Human Bcl-2 related protein A1 ELISA Kit (Colorimetric) measures human Bcl-2 related protein A1 in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 - 2000 pg/mL
This kit recognizes Human BCL2A1 in samples. No significant cross-reactivity or interference between Human BCL2A1 and analogues was observed.
Biological Fluids, Plasma, Serum
597
Human
Storage of components varies. See protocol for specific instructions.
18.75 pg/mL
ELISA Kit (Colorimetric)
ELISA
BCL2A1
1 Kit
Bcl-2 related protein A1
Product Suggestions
Product Suggestions