Learn More
BioworldSupplier Diversity Partner HUMAN BETA 2 MICROGLOBULIN DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220602531
HUMAN BETA 2 MICROGLOBULIN (RECOMBINANT): Recombinant Human Beta-2 Microglobulin (B2M) Source: E.Coli Background: An 11 kDa protein associated with the outer membrane of many cells including lymphocytes. It is the small subunit of th...
Catalog No. 50-253-351