Learn More
Novus Biologicals™ Human Beta-thromboglobulin ELISA Kit (Colorimetric)
Human Beta-thromboglobulin ELISA Kit (Colorimetric) measures Human Beta-thromboglobulin in serum, plasma, tissue homogenates
Supplier: Novus Biologicals™ NBP2888315KIT
Specifications
0.78 ng/mL-50 ng/mL | |
ELISA Kit (Colorimetric) | |
This assay has high sensitivity and excellent specificity for detection of human beta-TG. No significant cross-reactivity or interference between human beta-TG and analogues was observed. | |
ELISA | |
PPBP | |
Store at 4°C. | |
Beta-thromboglobulin |
0.195 ng/mL | |
Plasma, Serum, Tissue Homogenates | |
Human | |
5473 | |
5 Kit | |
Beta-TG, CTAP III, CXCL7, NAP-2, PBP, PPBP, TC1, TGB, THBGB1 |