Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Beta-thromboglobulin ELISA Kit (Colorimetric)
Human Beta-thromboglobulin ELISA Kit (Colorimetric) measures Human Beta-thromboglobulin in serum, plasma, tissue homogenates
$953.00 - $7244.00
Specifications
Assay Range | 0.78 ng/mL-50 ng/mL |
---|---|
Assay Sensitivity | 0.195 ng/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Serum, Plasma, Tissue Homogenates |
Specificity | This assay has high sensitivity and excellent specificity for detection of human beta-TG. No significant cross-reactivity or interference between human beta-TG and analogues was observed. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB036451
|
Novus Biologicals™
NBP28883110KIT |
10 Kit |
Each for $7,244.00
|
|
NB036452
|
Novus Biologicals™
NBP2888311KIT |
1 Kit |
Each for $953.00
|
|
NB036453
|
Novus Biologicals™
NBP2888315KIT |
5 Kit |
Each for $3,794.00
|
|
Specifications
0.78 ng/mL-50 ng/mL | |
ELISA Kit (Colorimetric) | |
This assay has high sensitivity and excellent specificity for detection of human beta-TG. No significant cross-reactivity or interference between human beta-TG and analogues was observed. | |
ELISA | |
PPBP | |
Beta-TG, CTAP III, CXCL7, NAP-2, PBP, PPBP, TC1, TGB, THBGB1 |
0.195 ng/mL | |
Serum, Plasma, Tissue Homogenates | |
Human | |
5473 | |
Store at 4°C. | |
Beta-thromboglobulin |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title