Learn More
Raybiotech IncSupplier Diversity Partner Human bFGF ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHBFGFCL1
For cell and tissue lysate samples. Sensitivity: 50 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human bFGF.
Catalog No. 50-194-9018