Learn More
Sigma Aldrich Fine Chemicals Biosciences Human bfGf ELISA Kit for s
Supplier:  Sigma Aldrich Fine Chemicals Biosciences RAB01821KT
Encompass
The Human bfGf ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human bfGf in serum plasma cell culture supernatants and urine.
Catalog No. 50-290-4941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.