Learn More
Novus Biologicals™ Human BLNK ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP269972
Catalog No. NBP269972
Add to cart
Description
Description
The Human BLNK ELISA Kit (Colorimetric) measures human BLNK in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human BLNK in samples. No significant cross-reactivity or interference between Human BLNK and analogues was observed.
29760
0.10 ng/mL
BLNK
BLNK
Biological Fluids, Plasma, Serum