Learn More
Novus Biologicals™ Human BMP-10 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269981
Catalog No. NBP269981
Add to Cart
Description
Description
The Human BMP-10 ELISA Kit (Colorimetric) measures human BMP-10 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Human BMP-10 in samples. No significant cross-reactivity or interference between Human BMP-10 and analogues was observed.
Biological Fluids, Plasma, Serum
27302
Human
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
BMP10
1 Kit
BMP-10
Recommendations
Recommendations