Learn More
Novus Biologicals™ Human BMP-6 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP269997
Catalog No. NBP269997
Add to cart
Description
Description
The Human BMP-6 ELISA Kit (Colorimetric) measures human BMP-6 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human BMP-6 in samples. No significant cross-reactivity or interference between Human BMP-6 and analogues was observed.
654
0.10 ng/mL
BMP-6
BMP6
Biological Fluids, Plasma, Serum