Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Brevican ELISA Kit Non-distribution product as customer accommodation.

Human Brevican quantitates human Brevican in serum, plasma, supernatant.

$491.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 0.041 ng/mL
Assay Range 0.041 to 10 ng/mL
Test Time 1 hr 20 min
Sample Volume Serum, 50 µL; Plasma, 50 µL; Supernatant, 50 µL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EH52RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
EH52RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Human Brevican quantitates human Brevican in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Brevican.

Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
Specifications

Specifications

2°C to 8°C
0.041 to 10 ng/mL
Serum, 50 µL; Plasma, 50 µL; Supernatant, 50 µL
<10%
0.041 ng/mL
1 hr 20 min
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit