Learn More
Abnova™ Human BTBD1 Partial ORF (NP_079514, 384 a.a. - 482 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_079514 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53339 |
Molecular Weight (g/mol) | 36.63kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-936-942
|
Abnova™
H00053339Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-936-943
|
Abnova™
H00053339Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYTSpecifications
NP_079514 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C15orf1/NS5ATP8 | |
BTBD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
53339 | |
BTBD1 (Human) Recombinant Protein (Q01) | |
EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYT | |
RUO | |
BTBD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.