Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human BTNL8 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9359N100UG
This item is not returnable.
View return policy
Description
BTNL8 Monoclonal specifically detects BTNL8 in Human samples. It is validated for Flow Cytometry.Specifications
BTNL8 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
BTN9.2, butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347 | |
Human embryonic kidney cell line HEK-293-derived Human BTNL8 Gln18-Lys238 Accession # Q6UX41-1 | |
100 μg | |
79908 | |
Human | |
Supernatant |
Flow Cytometry | |
2187b | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6UX41-1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human BTNL8 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction