Learn More
Abnova™ Human BVES Full-length ORF (NP_009004.2, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011149P01S
Description
This gene encodes a member of the POP family of proteins containing three putative transmembrane domains. This gene is expressed in cardiac and skeletal muscle and may play an important role in development of these tissues. The mouse ortholog may be involved in the regeneration of adult skeletal muscle and may act as a cell adhesion molecule in coronary vasculogenesis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: MNYTESSPLRESTAIGFTPELESIIPVPSNKTTCENWREIHHLVFHVANICFAVGLVIPTTLHLHMIFLRGMLTLGCTLYIVWATLYRCALDIMIWNSVFLGVNILHLSYLLYKKRPVKIEKELSGMYRRLFEPLRVPPDLFRRLTGQFCMIQTLKKGQTYAAEDKTSVDDRLSILLKGKMKVSYRGHFLHNIYPCAFIDSPEFRSTQMHKGEKFQVTIIADDNCRFLCWSRERLTYFLESEPFLYEIFRYLIGKDITNKLYSLNDPTLNDKKAKKLEHQLSLCTQISMLEMRNSIASSSDSDDGLHQFLRGTSSMSSLHVSSPHQRASAKMKPIEEGAEDDDDVFEPASPNTLKVHQLPSpecifications
NP_009004.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BVES | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
11149 | |
BVES (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBVES/MGC42413/POP1/POPDC1 | |
BVES | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.