Learn More
R&D Systems™ Human C-Reactive Protein/CRP Luminex Performance Assay
Fluorokine MAP for use in the Obesity Panel
Manufacturer:  R&D Systems™ LOB1707
Catalog No. LOB1707
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • User selects specific analytes of interest
  • Requires small sample size
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
Specifications
Specifications
1.9pg/mL
1 Kit
32.00 - 23500pg/mL
1401
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Cell Culture, Plasma (EDTA, Heparin), Serum
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Polystyrene
Human
C-Reactive Protein/CRP
CRP
Bead-based Multiplex Assay for the Luminex™ Platform
Cell Culture Supernates (50μL),Serum (15μL),EDTA Plasma (15μL),Heparin Plasma (15μL)