Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C1D Partial ORF (NP_006324, 77 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010438Q01L

 View more versions of this product

Catalog No. 89-937-276


Add to cart

Description

Description

The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Several alternatively spliced transcript variants of this gene have been described, but the full length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: NPKEHPVKQELERIRVYMNRVKEITDKKKAGKLDRGAASRFVKNALWEPKSKNASKVANKGKSKS
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.89kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NPKEHPVKQELERIRVYMNRVKEITDKKKAGKLDRGAASRFVKNALWEPKSKNASKVANKGKSKS
RUO
C1D
Wheat Germ (in vitro)
GST
NP_006324
Liquid
10438
C1D (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC12261/MGC14659/SUNCOR
C1D
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit