Learn More
Novus Biologicals™ Human C1q ELISA Kit (Chemiluminescence)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266749
Catalog No. NBP266749
Add to Cart
Description
Description
The Human C1q ELISA Kit (Chemiluminescence) measures human C1q in serum, plasma and other biological fluids.
Specifications
Specifications
15.63-1000 pg/mL
This kit recognizes Human C1q in samples. No significant cross-reactivity or interference between Human C1q and analogues was observed.
Biological Fluids, Plasma, Serum
712
Human
Storage of components varies. See protocol for specific instructions.
9.38 pg/mL
ELISA Kit (Chemiluminescence)
ELISA
C1QA
1 Kit
C1q
Product Suggestions
Product Suggestions