Learn More
Novus Biologicals™ Human C1qTNF3/CORS26/CTRP3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266753
Catalog No. NBP266753
Description
Description
The Human C1qTNF3/CORS26/CTRP3 ELISA Kit (Colorimetric) measures human C1qTNF3/CORS26/CTRP3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.63 - 40 ng/mL
This kit recognizes Human C1QTNF3 in samples. No significant cross-reactivity or interference between Human C1QTNF3 and analogues was observed.
114899
Human
1 Kit
Storage is content dependent.
0.38 ng/mL
ELISA
C1QTNF3
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
C1qTNF3/CORS26/CTRP3