Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Human C5a Platinum ELISA Kit Non-distribution product as customer accommodation.

For the detection of human C5a in serum, plasma (EDTA, citrate, heparin) and cell culture supernatant

Manufacturer:  Invitrogen™ BMS2088

 View more versions of this product

Catalog No. 50-112-5291


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C5a

Description

Description

C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.
Specifications

Specifications

P01031
Biotin
727
Adhesive Plate Covers
96 tests
0.005 ng/mL
Sample volume 10μL for plasma and 5μL for serum (prediluted 1:100 for serum, 1:25 for plasma), 50μL for CCS
Human
1 hr 20 min
Human
complement component 5, C5a, C5D, C5b, ECLZB, CPAMD4
3.6%
0.078-5.0 ng/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Plasma,10 μL|Serum,5 μL|Supernatant,50 μL
2-8°C
4 hr 30 min
C5
4%
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit