Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human C6orf108 (NP_006434, 1 a.a. - 174 a.a.) Full-length Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P3411

Catalog No. 89-937-569


Add to cart

Description

Description

This gene was identified on the basis of its stimulation by c-Myc protein. The latter is a transcription factor that participates in the regulation of cell proliferation, differentiation, and apoptosis. The exact function of this gene is not known but studies in rat suggest a role in cellular proliferation and c-Myc-mediated transformation. Two alternative transcripts encoding different proteins have been described. [provided by RefSeq]

Sequence: MGSSHHHHHHSSGLVPRGSHMAAAMVPGRSESWERGEPGRPALYFCGSIRGGREDRTLYERIVSRLRRFGTVLTEHVAAAELGARGEEAAGGDRLIHEQDLEWLQQADVVVAEVTQPSLGVGYELGRAVAFNKRILCLFRPQSGRVLSAMIRGAADGSRFQVWDYEEGEVEALLDRYFEADPPGQVAASPDPTT
Specifications

Specifications

Escherichia coli expression system
1 mg/mL
Liquid
10591
C6orf108 (Human) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 15% SDS-PAGE
MGSSHHHHHHSSGLVPRGSHMAAAMVPGRSESWERGEPGRPALYFCGSIRGGREDRTLYERIVSRLRRFGTVLTEHVAAAELGARGEEAAGGDRLIHEQDLEWLQQADVVVAEVTQPSLGVGYELGRAVAFNKRILCLFRPQSGRVLSAMIRGAADGSRFQVWDYEEGEVEALLDRYFEADPPGQVAASPDPTT
RUO
C6orf108
Escherichia coli
His
NP_006434
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
21.2kDa
Escherichia coli expression system
>95% by SDS-PAGE
100 ug
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
RCL/RP3-330M21.3/dJ330M21.3
C6orf108
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit