Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CA8 Partial ORF (NP_004047, 40 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000767Q01L

 View more versions of this product

Catalog No. 89-937-753


Add to cart

Description

Description

The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form. [provided by RefSeq]

Sequence: VFPDANGEYQSPINLNSREARYDPSLLDVRLSPNYVVCRDCEVTNDGHTIQVILKSKSVLSGGPLPQGHEFELYEVRFHWGRENQRGSEHTVNFKAFPME
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VFPDANGEYQSPINLNSREARYDPSLLDVRLSPNYVVCRDCEVTNDGHTIQVILKSKSVLSGGPLPQGHEFELYEVRFHWGRENQRGSEHTVNFKAFPME
RUO
CA8
Wheat Germ (in vitro)
GST
NP_004047
Liquid
767
CA8 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CA-VIII/CALS/CARP/MGC120502/MGC99509
CA8
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit