Learn More
Abnova™ Human CABIN1 Partial ORF (NP_036427, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_036427 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23523 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-937-765
|
Abnova™
H00023523Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-937-764
|
Abnova™
H00023523Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. At least four alternatively spliced transcripts have been found for this gene, but the full-length nature of most of them has not been determined. [provided by RefSeq]
Sequence: MIRIAALNASSTIEDDHEGSFKSHKTQTKEAQEAEAFALYHKALDLQKHDRFEESAKAYHELLEASLLREAVSSGDEKEGLKHPGLILKYSTYKNLAQLAAQREDLETAMSpecifications
NP_036427 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAIN/KIAA0330/PPP3IN | |
CABIN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23523 | |
CABIN1 (Human) Recombinant Protein (Q01) | |
MIRIAALNASSTIEDDHEGSFKSHKTQTKEAQEAEAFALYHKALDLQKHDRFEESAKAYHELLEASLLREAVSSGDEKEGLKHPGLILKYSTYKNLAQLAAQREDLETAM | |
RUO | |
CABIN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.