Learn More
Abnova™ Human CACNG7 Partial ORF (NP_114102.2, 203 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00059284Q01L
Description
The mouse protein stargazin is one of five subunits comprising neuronal voltage-gated calcium channels. This subunit, gamma, is thought to stabilize the calcium channel in an inactive (closed) state. Mutations in the gene encoding stargazin have been associated with absence seizures, also known as petit-mal or spike-wave seizures. The protein encoded by this gene is structurally similar to the mouse stargazin protein and is a member of the neuronal calcium channel gamma subunit protein family. However, it appears unlikely that the encoded protein is part of a functional calcium channel. Rather, it appears to inhibit the expression of a specific calcium channel subunit. [provided by RefSeq]
Sequence: RYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSPSpecifications
NP_114102.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYAEEEMYRPHPAFYRPRLSDCSDYSGQFLQPEAWRRGRSPSDISSDVSIQMTQNYPPAIKYPDHLHISTSP | |
RUO | |
CACNG7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
59284 | |
CACNG7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CACNG7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.