Learn More
Novus Biologicals™ Human Cadherin-19 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275168
Catalog No. NBP275168
Add to cart
Description
Description
The Human Cadherin-19 ELISA Kit (Colorimetric) measures human Cadherin-19 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CDH19 in samples. No significant cross-reactivity or interference between Human CDH19 and analogues was observed.
28513
0.10 ng/mL
Cadherin-19
CDH19
Biological Fluids, Plasma, Serum