Learn More
Novus Biologicals™ Human CGRP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275259
Catalog No. NBP275259
Add to cart
Description
Description
The Human Calcitonin ELISA Kit (Colorimetric) measures human Calcitonin in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP-I
Biological Fluids, Plasma, Serum
This kit recognizes Human CGRP1 in samples. No significant cross-reactivity or interference between Human CGRP1 and analogues was observed.
796
9.38 pg/mL
CGRP
CALCA
1 Kit