Learn More
BioworldSupplier Diversity Partner HUMAN CALPAIN-1 CATALYTIC S DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600001
HUMAN CALPAIN-1 CATALYTIC SUBUNIT: Calpain-1 (mu-Calpain) Source: Human Eryhtrocytes Description: This protein consists of an 80-kDa large subunit and a 30 kDa small subunit. The protein was purified by sequential chromatography t...
Catalog No. 50-253-093