Learn More
Novus Biologicals™ Human Calponin 1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275789
Catalog No. NBP275789
Add to cart
Description
Description
The Human Calponin 1 ELISA Kit (Colorimetric) measures human Calponin 1 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CNN1 in samples. No significant cross-reactivity or interference between Human CNN1 and analogues was observed.
1264
0.19 ng/mL
Calponin 1
CNN1
Biological Fluids, Plasma, Serum