Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CAPN10 Full-length ORF (NP_075571.1, 1 a.a. - 672 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011132P01

Catalog No. 89-937-953


Add to cart

Description

Description

Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large catalytic subunit has four domains: domain I, the N-terminal regulatory domain that is processed upon calpain activation; domain II, the protease domain; domain III, a linker domain of unknown function; and domain IV, the calmodulin-like calcium-binding domain. This gene encodes a large subunit. It is an atypical calpain in that it lacks the calmodulin-like calcium-binding domain and instead has a divergent C-terminal domain. It is similar in organization to calpains 5 and 6. This gene is associated with type 2 or non-insulin-dependent diabetes mellitus (NIDDM) and located within the NIDDM1 region. Multiple alternative transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MRAGRGATPARELFRDAAFPAADSSLFCDLSTPLAQFREDITWRRPQEICATPRLFPDDPREGQVKQGLLGDCWFLCACAALQKSRHLLDQVIPPGQPSWADQEYRGSFTCRIWQFGRWVEVTTDDRLPCLAGRLCFSRCQREDVFWLPLLEKVYAKVHGSYEHLWAGQVADALVDLTGGLAERWNLKGVAGSGGQQDRPGRWEHRTCRQLLHLKDQCLISCCVLSPRAGARELGEFHAFIVSDLRELQGQAGQCILLLRIQNPWGRRCWQGLWREGGEGWSQVDAAVASELLSQLQEGEFWVEEEEFLREFDELTVGYPVTEAGHLQSLYTERLLCHTRALPGAWVKGQSAGGCRNNSGFPSNPKFWLRVSEPSEVYIAVLQRSRLHAADWAGRARALVGDSHTSWSPASIPGKHYQAVGLHLWKVEKRRVNLPRVLSMPPVAGTACHAYDREVHLRCELSPGYYLAVPSTFLKDAPGEFLLRVFSTGRVSLSAIRAVAKNTTPGAALPAGEWGTVQLRGSWRVGQTAGGSRNFASYPTNPCFPFSVPEGPGPRCVRITLHQHCRPSDTEFHPIGFHIFQVPEGGRSQDAPPLLLQEPLLSCVPHRYAQEVSRLCLLPAGTYKVVPSTYLPDTEGAFTVTIATRIDRPSIHSQEMLGQFLQEVSVMAVMKT
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
101.3kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAPN10
Wheat Germ (in vitro)
GST
NP_075571.1
Liquid
11132
CAPN10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRAGRGATPARELFRDAAFPAADSSLFCDLSTPLAQFREDITWRRPQEICATPRLFPDDPREGQVKQGLLGDCWFLCACAALQKSRHLLDQVIPPGQPSWADQEYRGSFTCRIWQFGRWVEVTTDDRLPCLAGRLCFSRCQREDVFWLPLLEKVYAKVHGSYEHLWAGQVADALVDLTGGLAERWNLKGVAGSGGQQDRPGRWEHRTCRQLLHLKDQCLISCCVLSPRAGARELGEFHAFIVSDLRELQGQAGQCILLLRIQNPWGRRCWQGLWREGGEGWSQVDAAVASELLSQLQEGEFWVEEEEFLREFDELTVGYPVTEAGHLQSLYTERLLCHTRALPGAWVKGQSAGGCRNNSGFPSNPKFWLRVSEPSEVYIAVLQRSRLHAADWAGRARALVGDSHTSWSPASIPGKHYQAVGLHLWKVEKRRVNLPRVLSMPPVAGTACHAYDREVHLRCELSPGYYLAVPSTFLKDAPGEFLLRVFSTGRVSLSAIRAVAKNTTPGAALPAGEWGTVQLRGSWRVGQTAGGSRNFASYPTNPCFPFSVPEGPGPRCVRITLHQHCRPSDTEFHPIGFHIFQVPEGGRSQDAPPLLLQEPLLSCVPHRYAQEVSRLCLLPAGTYKVVPSTYLPDTEGAFTVTIATRIDRPSIHSQEMLGQFLQEVSVMAVMKT
RUO
CAPN10
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit