Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CAPN7 Partial ORF (NP_055111.1, 714 a.a. - 813 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023473Q01L

 View more versions of this product

Catalog No. 89-937-972


Add to cart

Description

Description

Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large subunit possesses a cysteine protease domain, and both subunits possess calcium-binding domains. Calpains have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. The function of the protein encoded by this gene is not known. An orthologue has been found in mouse but it seems to diverge from other family members. The mouse orthologue is thought to be calcium independent with protease activity. [provided by RefSeq]

Sequence: NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQ
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NPIYQFHIEKTGPLLIELRGPRQYSVGFEVVTVSTLGDPGPHGFLRKSSGDYRCGFCYLELENIPSGIFNIIPSTFLPKQEGPFFLDFNSIIPIKITQLQ
RUO
CAPN7
Wheat Germ (in vitro)
GST
NP_055111.1
Liquid
23473
CAPN7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CALPAIN7/FLJ36423/PALBH
CAPN7
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit