Learn More
BioworldSupplier Diversity Partner HUMAN CARBONIC ANHYDRASE II DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220602681
HUMAN CARBONIC ANHYDRASE III (RECOMBINANT): Recombinant Human Carbonic Human Anhydrase III Source:E.Coli Description: Recombinant Human CAIII, Carbonic anhydrase III having a Mw of 33.9kDa is one of a group of ten zinc metalloenzymes...
Catalog No. 50-253-366