Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbonic Anhydrase X/CA10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2189SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase X/CA10 Monoclonal specifically detects Carbonic Anhydrase X/CA10 in Human samples. It is validated for Western Blot.Specifications
Carbonic Anhydrase X/CA10 | |
Monoclonal | |
LYOPH | |
Western Blot 1.5 ug/mL | |
Q9NS85 | |
CA10 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase X/CA10 Met1-Asn300 Accession # Q9NS85 | |
25 μg | |
Primary | |
Detects human Carbonic Anhydrase X/CA10 in ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
862319 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA10, carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CARPX, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
56934 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction