Learn More
Novus Biologicals™ Human Carbonic Anhydrase XII/CA12 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP270053
Catalog No. NBP270053
Add to Cart
Description
Description
The Human Carbonic Anhydrase XII/CA12 ELISA Kit (Colorimetric) measures human Carbonic Anhydrase XII/CA12 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Human CA12 in samples. No significant cross-reactivity or interference between Human CA12 and analogues was observed.
Biological Fluids, Plasma, Serum
771
Human
Storage of components varies. See protocol for specific instructions.
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
CA12
1 Kit
Carbonic Anhydrase XII/CA12
Recommendations
Recommendations