Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CARD10 Partial ORF (NP_055365, 566 a.a. - 655 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00029775Q01L

 View more versions of this product

Catalog No. 89-938-007


Add to cart

Description

Description

The caspase recruitment domain (CARD) is a protein module that consists of 6 or 7 antiparallel alpha helices. It participates in apoptosis signaling through highly specific protein-protein homophilic interactions. Like several other CARD proteins, CARD10 belongs to the membrane-associated guanylate kinase (MAGUK) family and activates NF-kappa-B (NFKB; see MIM 164011) through BCL10 (MIM 603517) (Wang et al., 2001 [PubMed 11259443]).[supplied by OMIM]

Sequence: LSSSSSSDSVWPLGKPEGLLARGCGLDFLNRSLAIRVSGRSPPGGPEPQDKGPDGLSFYGDRWSGAVVRRVLSGPGSARMEPREQRVEAA
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LSSSSSSDSVWPLGKPEGLLARGCGLDFLNRSLAIRVSGRSPPGGPEPQDKGPDGLSFYGDRWSGAVVRRVLSGPGSARMEPREQRVEAA
RUO
CARD10
Wheat Germ (in vitro)
GST
NP_055365
Liquid
29775
CARD10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BIMP1/CARMA3/MGC142219
CARD10
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit