Learn More
Abnova™ Human CARD12 Partial ORF (AAH31555, 531 a.a. - 630 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00058484Q01L
Description
In C. elegans, Ced4 binds and activates Ced3, an apoptotic initiator caspase, via caspase-associated recruitment domains (CARDs). Human Ced4 homologs include APAF1 (MIM 602233), NOD1/CARD4 (MIM 605980), and NOD2/CARD15 (MIM 605956). These proteins have at least 1 N-terminal CARD domain followed by a centrally located nucleotide-binding domain (NBD or NACHT) and a C-terminal regulatory domain, found only in mammals, that contains either WD40 repeats or leucine-rich repeats (LRRs). CARD12 is a member of the Ced4 family and can induce apoptosis.[supplied by OMIM]
Sequence: QESLQSVKNTTEQEILKAININSFVECGIHLYQESTSKSALSQEFEAFFQGKSLYINSGNIPDYLFDFFEHLPNCASALDFIKLDFYGGAMASWEKAAEDSpecifications
AAH31555 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QESLQSVKNTTEQEILKAININSFVECGIHLYQESTSKSALSQEFEAFFQGKSLYINSGNIPDYLFDFFEHLPNCASALDFIKLDFYGGAMASWEKAAED | |
RUO | |
NLRC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
58484 | |
CARD12 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARD12/CLAN/CLAN1/CLANA/CLANB/CLANC/CLAND/CLR2.1/IPAF | |
NLRC4 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.