Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CARD14 Full-length ORF (NP_438170.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name CARD14 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 74.9kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-938-016
View Documents
Abnova™
H00079092P01S
10 ug
Each for $460.50
Add to cart
 
89-938-017
View Documents
Abnova™
H00079092P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: MVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFSSCLEIPEGDPGALPGAKAGDPHLDYELLDTADLPQLESSLQPVSPGRLDVSESGVLMRRRPARRILSQVTMLAFQGDALLEQISVIGGNLTGIFIHRVTPGSAADQMALRPGTQIVMVSRARPLLSPGLLMGTVAAGGVTQADFTSPRRCRSTLGWASALSWADVKRSAHL
Specifications

Specifications

CARD14 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
74.9kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit