Learn More
Abnova™ Human CARKL Full-length ORF (NP_037408.2, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023729P01S
Description
The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements. [provided by RefSeq]
Sequence: MAARPITLGIDLGTTSVKAALLRAAPDDPSGFAVLASCARAARAEAAVESAVAGPQGREQDVSRILQALHECLAALPRPQLRSVVGIGVSGQMHGVVFWKTGQGCEWTEGGITPVFEPRAVSHLVTWQDGRCSSEFLASLPQPKSHLSVATGFGCATIFWLLKYRPEFLKSYDAAGTIHDYVVAMLCGLPRPLMSDQNAASWGYFNTQSQSWNVETLRSSGFPVHLLPDIAEPGSVAGRTSHMWFEIPKGTQVGVALGDLQASVYSCMAQRTDAVLNISTSVQLAASMPSGFQPAQTPDPTAPVAYFPYFNRTYLGVAASLNGGNVLATFVHMLVQWMADLGLEVEESTVYSRMIQAAVQQRDTHLTITPTVLGERHLPDQLASVTRISSSDLSLGHVTRALCRGIVQNLHSMLPIQQLQEWGVERVMGSGSALSRNDVLKQEVQRAFPLPMSFGQDVDAAVGAALVMLRRHLNQKESSpecifications
NP_037408.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARKL/FLJ32478/SHK | |
SHPK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23729 | |
CARKL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAARPITLGIDLGTTSVKAALLRAAPDDPSGFAVLASCARAARAEAAVESAVAGPQGREQDVSRILQALHECLAALPRPQLRSVVGIGVSGQMHGVVFWKTGQGCEWTEGGITPVFEPRAVSHLVTWQDGRCSSEFLASLPQPKSHLSVATGFGCATIFWLLKYRPEFLKSYDAAGTIHDYVVAMLCGLPRPLMSDQNAASWGYFNTQSQSWNVETLRSSGFPVHLLPDIAEPGSVAGRTSHMWFEIPKGTQVGVALGDLQASVYSCMAQRTDAVLNISTSVQLAASMPSGFQPAQTPDPTAPVAYFPYFNRTYLGVAASLNGGNVLATFVHMLVQWMADLGLEVEESTVYSRMIQAAVQQRDTHLTITPTVLGERHLPDQLASVTRISSSDLSLGHVTRALCRGIVQNLHSMLPIQQLQEWGVERVMGSGSALSRNDVLKQEVQRAFPLPMSFGQDVDAAVGAALVMLRRHLNQKES | |
RUO | |
SHPK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.