Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CARKL Full-length ORF (NP_037408.2, 1 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023729P01L

 View more versions of this product

Catalog No. 89-938-038


Add to cart

Description

Description

The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements. [provided by RefSeq]

Sequence: MAARPITLGIDLGTTSVKAALLRAAPDDPSGFAVLASCARAARAEAAVESAVAGPQGREQDVSRILQALHECLAALPRPQLRSVVGIGVSGQMHGVVFWKTGQGCEWTEGGITPVFEPRAVSHLVTWQDGRCSSEFLASLPQPKSHLSVATGFGCATIFWLLKYRPEFLKSYDAAGTIHDYVVAMLCGLPRPLMSDQNAASWGYFNTQSQSWNVETLRSSGFPVHLLPDIAEPGSVAGRTSHMWFEIPKGTQVGVALGDLQASVYSCMAQRTDAVLNISTSVQLAASMPSGFQPAQTPDPTAPVAYFPYFNRTYLGVAASLNGGNVLATFVHMLVQWMADLGLEVEESTVYSRMIQAAVQQRDTHLTITPTVLGERHLPDQLASVTRISSSDLSLGHVTRALCRGIVQNLHSMLPIQQLQEWGVERVMGSGSALSRNDVLKQEVQRAFPLPMSFGQDVDAAVGAALVMLRRHLNQKES
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CARKL/FLJ32478/SHK
SHPK
Yes
NP_037408.2
Liquid
23729
CARKL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAARPITLGIDLGTTSVKAALLRAAPDDPSGFAVLASCARAARAEAAVESAVAGPQGREQDVSRILQALHECLAALPRPQLRSVVGIGVSGQMHGVVFWKTGQGCEWTEGGITPVFEPRAVSHLVTWQDGRCSSEFLASLPQPKSHLSVATGFGCATIFWLLKYRPEFLKSYDAAGTIHDYVVAMLCGLPRPLMSDQNAASWGYFNTQSQSWNVETLRSSGFPVHLLPDIAEPGSVAGRTSHMWFEIPKGTQVGVALGDLQASVYSCMAQRTDAVLNISTSVQLAASMPSGFQPAQTPDPTAPVAYFPYFNRTYLGVAASLNGGNVLATFVHMLVQWMADLGLEVEESTVYSRMIQAAVQQRDTHLTITPTVLGERHLPDQLASVTRISSSDLSLGHVTRALCRGIVQNLHSMLPIQQLQEWGVERVMGSGSALSRNDVLKQEVQRAFPLPMSFGQDVDAAVGAALVMLRRHLNQKES
RUO
SHPK
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit