Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human CASC5 Partial ORF (NP_733468, 3 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00057082Q01L
Description
Sequence: GVSSEANEENDNIERPVRRRHSSILKPPRSPLQDHRGGNERVQESNALRNKKNSRRVSFADTIKVFQTESHMKIVRKSEMEGCSAMVPSQLQLLPPSpecifications
NP_733468 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GVSSEANEENDNIERPVRRRHSSILKPPRSPLQDHRGGNERVQESNALRNKKNSRRVSFADTIKVFQTESHMKIVRKSEMEGCSAMVPSQLQLLPP | |
RUO | |
CASC5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57082 | |
CASC5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AF15Q14/D40/KIAA1570/KNL1 | |
CASC5 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction