Learn More
Raybiotech IncSupplier Diversity Partner Human Caspase-3 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHCASP31
For Serum, Plasma, and Cell Culture Supernatants. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Fes.
Catalog No. 50-194-9059